Product Details

SNP ID
rs7893132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:22359151 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTATATTATTTCTTATTGGACCC[C/G]AGAAAGAATGCCTGAGTCAGATGGT
Phenotype
MIM: 605730
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPAG6 PubMed Links

Gene Details

Gene
SPAG6
Gene Name
sperm associated antigen 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253854.1 Intron NP_001240783.1
NM_001253855.1 Intron NP_001240784.1
NM_012443.3 Intron NP_036575.1
NM_172242.2 Intron NP_758442.1
XM_005252645.1 Intron XP_005252702.1
XM_005252646.1 Intron XP_005252703.1

View Full Product Details