Product Details

SNP ID
rs3815528
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:12332175 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTATGTGTGGGAGACAAAGAAATA[C/T]CGAAAGCAAGTAAAGAAAAAAAAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VWDE PubMed Links

Gene Details

Gene
VWDE
Gene Name
von Willebrand factor D and EGF domains
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135924.1 Intron NP_001129396.1
XM_005249641.1 Intron XP_005249698.1
XM_011515174.1 Intron XP_011513476.1
XM_011515175.1 Intron XP_011513477.1
XM_017011828.1 Intron XP_016867317.1
XM_017011829.1 Intron XP_016867318.1
XM_017011830.1 Intron XP_016867319.1

View Full Product Details