Product Details

SNP ID
rs2722190
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:103950416 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCATAGGGAGTTGTGGTGGGCCCA[G/T]CAGAGTTGCCCACCACTCACGTGCT
Phenotype
MIM: 191175
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C12orf73 PubMed Links

Gene Details

Gene
C12orf73
Gene Name
chromosome 12 open reading frame 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135570.1 1543 UTR 3 NP_001129042.1
XM_011538718.2 1543 UTR 3 XP_011537020.1
XM_017019916.1 1543 Intron XP_016875405.1
XM_017019917.1 1543 UTR 3 XP_016875406.1
Gene
HSP90B1
Gene Name
heat shock protein 90 beta family member 1
There are no transcripts associated with this gene.

View Full Product Details