Product Details
- SNP ID
-
rs7303338
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:108590925 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGCAGTTTCAAAATACTGCTTCAC[G/T]CCAGGCTGCATGTTGCCTTACGAAC
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM119
PubMed Links
Gene Details
- Gene
- TMEM119
- Gene Name
- transmembrane protein 119
View Full Product Details