Product Details

SNP ID
rs7853557
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:38408691 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTATTTTCCCAAAAGGCCATCTTA[A/C]TCTAGGATGGCAGGGCTAAAAGGGA
Phenotype
MIM: 610413
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IGFBPL1 PubMed Links

Gene Details

Gene
IGFBPL1
Gene Name
insulin like growth factor binding protein like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007563.2 1396 UTR 3 NP_001007564.1
XM_017014699.1 1396 Intron XP_016870188.1

View Full Product Details