Product Details

SNP ID
rs7213637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38430232 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTCACAGAGAGGGTCTGAGCCTA[C/T]GTAGTGACAGTGCCCAGGCAAGAAC
Phenotype
MIM: 610590
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP23 PubMed Links

Gene Details

Gene
ARHGAP23
Gene Name
Rho GTPase activating protein 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199417.1 Intron NP_001186346.1
XM_006721991.1 Intron XP_006722054.1
XM_006721992.3 Intron XP_006722055.1
XM_006721993.3 Intron XP_006722056.1
XM_006721994.3 Intron XP_006722057.1
XM_011525073.1 Intron XP_011523375.1
XM_011525074.2 Intron XP_011523376.1
XM_011525075.2 Intron XP_011523377.1
XM_011525076.2 Intron XP_011523378.1
XM_011525077.2 Intron XP_011523379.1
XM_017024902.1 Intron XP_016880391.1

View Full Product Details