Product Details

SNP ID
rs7083576
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:95038717 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGTTAACTCCTGCAAGCCCCGCT[A/G]TCTGGCCCAATAATGATGGTGTATT
Phenotype
MIM: 601129
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYP2C8 PubMed Links

Gene Details

Gene
CYP2C8
Gene Name
cytochrome P450 family 2 subfamily C member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000770.3 Intron NP_000761.3
NM_001198853.1 Intron NP_001185782.1
NM_001198854.1 Intron NP_001185783.1
NM_001198855.1 Intron NP_001185784.1

View Full Product Details