Product Details

SNP ID
rs6860855
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:171434350 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATACCTGTGGGCCCAGTCCAGGAT[C/T]GGGAGATACATGCAGGTCAGCTTGG
Phenotype
MIM: 603726
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGF18 PubMed Links

Gene Details

Gene
FGF18
Gene Name
fibroblast growth factor 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003862.2 Intron NP_003853.1

View Full Product Details