Product Details

SNP ID
rs8113269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55486963 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCAGCAAAGACCTGTGAAGCTAC[A/G]GACTGACAGCCAGGGCAGGGGAGGA
Phenotype
MIM: 610671
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAT14 PubMed Links

Gene Details

Gene
NAT14
Gene Name
N-acetyltransferase 14 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020378.3 745 UTR 3 NP_065111.1
Gene
SSC5D
Gene Name
scavenger receptor cysteine rich family member with 5 domains
There are no transcripts associated with this gene.

Gene
ZNF628
Gene Name
zinc finger protein 628
There are no transcripts associated with this gene.

View Full Product Details