Product Details
- SNP ID
-
rs7110062
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:74180456 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTATTTCCCCATCAGTTCCCTTAA[C/G]ATGGTATAACTAGGGCTATACTGCC
- Phenotype
-
MIM: 615944
MIM: 611117
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C2CD3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80163117] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C2CD3
- Gene Name
- C2 calcium dependent domain containing 3
There are no transcripts associated with this gene.
- Gene
- PPME1
- Gene Name
- protein phosphatase methylesterase 1
View Full Product Details