Product Details
- SNP ID
-
rs6513496
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:60308476 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGACAGTTCAGAAGCCTGGAGA[C/T]CAGGAGTCTGCCAGTGGAGTCAGCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR646
PubMed Links
Gene Details
- Gene
- MIR646
- Gene Name
- microRNA 646
There are no transcripts associated with this gene.
- Gene
- MIR646HG
- Gene Name
- MIR646 host gene
There are no transcripts associated with this gene.
View Full Product Details