Product Details

SNP ID
rs7811571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103472661 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTGTCATTAGTTCACAGTGTGGG[A/G]AAGTGGTGTACACTCGGTCTTGAGA
Phenotype
MIM: 600514
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927870 PubMed Links

Gene Details

Gene
LOC101927870
Gene Name
uncharacterized LOC101927870
There are no transcripts associated with this gene.

Gene
RELN
Gene Name
reelin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005045.3 10694 UTR 3 NP_005036.2
NM_173054.2 10694 UTR 3 NP_774959.1

View Full Product Details