Product Details

SNP ID
rs7825412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:64374168 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGACTGAATGAGGGTGGGAGAGAG[A/G]GTGTGTCTCACGGCGCCCAGGAGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC102724623 PubMed Links

Gene Details

Gene
LOC102724623
Gene Name
uncharacterized LOC102724623
There are no transcripts associated with this gene.

Gene
MIR124-2
Gene Name
microRNA 124-2
There are no transcripts associated with this gene.

Gene
MIR124-2HG
Gene Name
MIR124-2 host gene
There are no transcripts associated with this gene.

View Full Product Details