Product Details

SNP ID
rs7871609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114787155 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCAAATAATAACCTGTCTCTACA[C/T]GTAGTTTAAATAATGGATTCTACAA
Phenotype
MIM: 604052
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFSF15 PubMed Links

Gene Details

Gene
TNFSF15
Gene Name
tumor necrosis factor superfamily member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204344.1 4169 UTR 3 NP_001191273.1
NM_005118.3 4169 UTR 3 NP_005109.2

View Full Product Details