Product Details

SNP ID
rs3764237
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34083969 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTTCAAGCCTCAGATAATTGAAG[A/T]AAATTGTCAGAAAAAAAGAAACATT
Phenotype
MIM: 605265
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
AVEN PubMed Links
Additional Information
For this assay, SNP(s) [rs138487396] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
AVEN
Gene Name
apoptosis and caspase activation inhibitor
There are no transcripts associated with this gene.

Gene
EMC7
Gene Name
ER membrane protein complex subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020154.2 Intron NP_064539.1

View Full Product Details