Product Details

SNP ID
rs7782241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:38181960 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTGTCCCTCTAAGTCCACTCAAA[G/T]GCTGCTTTATCAATAAAGCATTTCC
Phenotype
MIM: 611759
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
STARD3NL PubMed Links

Gene Details

Gene
STARD3NL
Gene Name
STARD3 N-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032016.3 Intron NP_114405.1
XM_005249880.1 Intron XP_005249937.1
XM_005249881.2 Intron XP_005249938.1
XM_011515572.1 Intron XP_011513874.1
XM_017012692.1 Intron XP_016868181.1
XM_017012693.1 Intron XP_016868182.1
XM_017012694.1 Intron XP_016868183.1
XM_017012695.1 Intron XP_016868184.1
XM_017012696.1 Intron XP_016868185.1
XM_017012697.1 Intron XP_016868186.1
XM_017012698.1 Intron XP_016868187.1

View Full Product Details