Product Details

SNP ID
rs7882031
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:151954019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCAGACGAAAGATGGAGCCAGCA[A/T]GGAGAGGGAGATGTCACAGCAGCAG
Phenotype
MIM: 300093 MIM: 300769
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GABRE PubMed Links

Gene Details

Gene
GABRE
Gene Name
gamma-aminobutyric acid type A receptor epsilon subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004961.3 1641 UTR 3 NP_004952.2
XM_011531140.2 1641 UTR 3 XP_011529442.1
XM_017029386.1 1641 UTR 3 XP_016884875.1
XM_017029387.1 1641 UTR 3 XP_016884876.1
XM_017029388.1 1641 UTR 3 XP_016884877.1
XM_017029389.1 1641 UTR 3 XP_016884878.1
Gene
MIR224
Gene Name
microRNA 224
There are no transcripts associated with this gene.

Gene
MIR452
Gene Name
microRNA 452
There are no transcripts associated with this gene.

View Full Product Details