Product Details

SNP ID
rs8081992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:21240908 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCTGGAGACCTTGGGCCAGGCCCC[A/G]CGAAGCCCTGCCTAGCCCCTTGGCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NATD1 PubMed Links

Gene Details

Gene
NATD1
Gene Name
N-acetyltransferase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152914.2 2749 UTR 3 NP_690878.2

View Full Product Details