Product Details
- SNP ID
-
rs7671851
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:165086117 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTATGGAAAGTACCTTCAATAGAAG[A/C]TTGTGCAAGAGGCTTGCACAAGGAG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM192
PubMed Links
Gene Details
- Gene
- TMEM192
- Gene Name
- transmembrane protein 192
View Full Product Details