Product Details

SNP ID
rs7769892
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159172113 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGATGCCACAGTAAAATTATTAA[C/T]TTTACCAAATCTTTGTATCTTTGAG
Phenotype
MIM: 609991
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FNDC1 PubMed Links

Gene Details

Gene
FNDC1
Gene Name
fibronectin type III domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032532.2 Intron NP_115921.2
XM_011536190.2 Intron XP_011534492.1
XM_011536191.2 Intron XP_011534493.1
Gene
LOC101929122
Gene Name
uncharacterized LOC101929122
There are no transcripts associated with this gene.

View Full Product Details