Product Details

SNP ID
rs6921032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:17401600 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTGATTGATGGACAGTTTCAACT[A/G]ATTGGTTACTTGACATGAGGTAGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CAP2 PubMed Links

Gene Details

Gene
CAP2
Gene Name
CAP, adenylate cyclase-associated protein, 2 (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006366.2 Intron NP_006357.1
XM_011514233.2 Intron XP_011512535.1

View Full Product Details