Product Details

SNP ID
rs7100410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:132331649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCGAGGACCGCTCCAAAGGTGGT[A/G]CCTCAGCAGCAATTCTTTTCTGCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC27 PubMed Links

Gene Details

Gene
LRRC27
Gene Name
leucine rich repeat containing 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143757.1 245 Intron NP_001137229.1
NM_001143758.1 245 Intron NP_001137230.1
NM_001143759.1 245 Intron NP_001137231.1
NM_001309474.1 245 Intron NP_001296403.1
NM_030626.2 245 Intron NP_085129.1
XM_006717983.2 245 Intron XP_006718046.1
XM_006717984.2 245 Intron XP_006718047.1
XM_011540208.1 245 Intron XP_011538510.1
XM_011540209.1 245 Intron XP_011538511.1
XM_011540210.2 245 Intron XP_011538512.1
XM_017016690.1 245 Intron XP_016872179.1
XM_017016691.1 245 Intron XP_016872180.1
XM_017016692.1 245 Intron XP_016872181.1
XM_017016693.1 245 Intron XP_016872182.1
XM_017016694.1 245 Intron XP_016872183.1
Gene
STK32C
Gene Name
serine/threonine kinase 32C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318878.1 245 Missense Mutation CAC,TAC H30Y NP_001305807.1
NM_001318879.1 245 Intron NP_001305808.1
NM_001318880.1 245 Missense Mutation CAC,TAC H30Y NP_001305809.1
NM_001318881.1 245 Missense Mutation CAC,TAC H30Y NP_001305810.1
NM_001318882.1 245 Missense Mutation CAC,TAC H30Y NP_001305811.1
NM_001318883.1 245 Missense Mutation CAC,TAC H30Y NP_001305812.1
NM_173575.3 245 Intron NP_775846.2
XM_011539688.1 245 Missense Mutation CAC,TAC H30Y XP_011537990.1
XM_011539690.2 245 Intron XP_011537992.1
XM_011539693.2 245 Intron XP_011537995.1
XM_011539694.1 245 Intron XP_011537996.1
XM_011539695.1 245 Intron XP_011537997.1
XM_011539696.1 245 Intron XP_011537998.1
XM_017016100.1 245 Missense Mutation CAC,TAC H30Y XP_016871589.1
XM_017016101.1 245 Missense Mutation CAC,TAC H30Y XP_016871590.1

View Full Product Details