Product Details

SNP ID
rs7896688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:92452139 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTGCTTTCTGCATTATGAACAAT[A/G]AAAGCAAGAAATCTGGTCAAACTGG
Phenotype
MIM: 146680
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IDE PubMed Links

Gene Details

Gene
IDE
Gene Name
insulin degrading enzyme
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165946.1 4025 UTR 3 NP_001159418.1
NM_001322793.1 4025 UTR 3 NP_001309722.1
NM_001322794.1 4025 UTR 3 NP_001309723.1
NM_001322795.1 4025 UTR 3 NP_001309724.1
NM_001322796.1 4025 UTR 3 NP_001309725.1
NM_001322797.1 4025 UTR 3 NP_001309726.1
NM_004969.3 4025 UTR 3 NP_004960.2
XM_017016187.1 4025 Intron XP_016871676.1
XM_017016188.1 4025 Intron XP_016871677.1
XM_017016189.1 4025 Intron XP_016871678.1
XM_017016190.1 4025 Intron XP_016871679.1

View Full Product Details