Product Details

SNP ID
rs7155258
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:22989505 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTGCATAAACAGAATGCATGATG[C/G]TGTTCTAGCCCTTTCCTTCTCTCAT
Phenotype
MIM: 609066
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AJUBA PubMed Links

Gene Details

Gene
AJUBA
Gene Name
ajuba LIM protein
There are no transcripts associated with this gene.

Gene
C14orf93
Gene Name
chromosome 14 open reading frame 93
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130706.2 Intron NP_001124178.1
NM_001130708.2 Intron NP_001124180.1
NM_001282968.1 Intron NP_001269897.1
NM_001282969.1 Intron NP_001269898.1
NM_001282970.1 Intron NP_001269899.1
NM_021944.3 Intron NP_068763.2
XM_005267971.1 Intron XP_005268028.1
XM_005267972.1 Intron XP_005268029.1
XM_006720231.1 Intron XP_006720294.1
XM_006720232.2 Intron XP_006720295.1
XM_011537059.1 Intron XP_011535361.1
XM_011537060.2 Intron XP_011535362.1
XM_011537062.1 Intron XP_011535364.1
XM_017021569.1 Intron XP_016877058.1
XM_017021570.1 Intron XP_016877059.1
XM_017021571.1 Intron XP_016877060.1
XM_017021572.1 Intron XP_016877061.1
XM_017021573.1 Intron XP_016877062.1
XM_017021574.1 Intron XP_016877063.1
XM_017021575.1 Intron XP_016877064.1
XM_017021576.1 Intron XP_016877065.1
XM_017021577.1 Intron XP_016877066.1

View Full Product Details