Product Details

SNP ID
rs8004860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23941591 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAATGGGAACTTATATTAGGCAT[C/G]CTATGCCTGGTGGATTTGTCCCTCT
Phenotype
MIM: 616925
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DHRS4-AS1 PubMed Links

Gene Details

Gene
DHRS4-AS1
Gene Name
DHRS4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details