Product Details

SNP ID
rs7128270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112248929 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTGTAATCATGAAGAAAGGCTTG[A/G]AGGCTGAGCTCTGGGGAATCTTGGC
Phenotype
MIM: 611904
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLET1 PubMed Links

Gene Details

Gene
PLET1
Gene Name
placenta expressed transcript 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145024.1 765 Missense Mutation TCC,TTC S165F NP_001138496.1

View Full Product Details