Product Details

SNP ID
rs8135102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42797122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATAAATGAAGCAATCTGTCACA[C/T]GAGACACCAGTCACTTTTGGTCTTT
Phenotype
MIM: 612439
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARFGAP3 PubMed Links

Gene Details

Gene
ARFGAP3
Gene Name
ADP ribosylation factor GTPase activating protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142293.1 2237 UTR 3 NP_001135765.1
NM_014570.4 2237 UTR 3 NP_055385.3
XM_005261525.3 2237 UTR 3 XP_005261582.1

View Full Product Details