Product Details

SNP ID
rs7271505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62145908 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTGTGTGCTGGTAGTGTCATGCC[A/G]AGAGCTTTAAGAGAAGCCAGGGACC
Phenotype
MIM: 606607 MIM: 606472
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PSMA7 PubMed Links

Gene Details

Gene
PSMA7
Gene Name
proteasome subunit alpha 7
There are no transcripts associated with this gene.

Gene
SS18L1
Gene Name
SS18L1, nBAF chromatin remodeling complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301778.1 Intron NP_001288707.1
NM_198935.2 Intron NP_945173.1
XM_005260389.3 Intron XP_005260446.1
XM_005260391.1 Intron XP_005260448.1
XM_011528764.2 Intron XP_011527066.1
XM_011528767.1 Intron XP_011527069.1
XM_017027784.1 Intron XP_016883273.1

View Full Product Details