Product Details

SNP ID
rs8120120
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.20:61975267 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAATTATAGTTTACATAGCCATTT[C/T]TATGTCATGTGCCAAAAGTTATGTA
Phenotype
MIM: 601796
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TAF4 PubMed Links

Gene Details

Gene
TAF4
Gene Name
TATA-box binding protein associated factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003185.3 4159 UTR 3 NP_003176.2

View Full Product Details