Product Details

SNP ID
rs8040730
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:51448192 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTATTGGTGTAGTTTCAACATA[C/T]CCTTAAATGTTTGGGTGTTTAACCT
Phenotype
MIM: 612186
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DMXL2 PubMed Links

Gene Details

Gene
DMXL2
Gene Name
Dmx like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174116.1 10254 UTR 3 NP_001167587.1
NM_001174117.1 10254 UTR 3 NP_001167588.1
NM_015263.3 10254 UTR 3 NP_056078.2
XM_005254255.1 10254 UTR 3 XP_005254312.1
XM_005254256.1 10254 UTR 3 XP_005254313.1
XM_011521402.2 10254 Intron XP_011519704.1
XM_017022034.1 10254 UTR 3 XP_016877523.1

View Full Product Details