Product Details

SNP ID
rs8035628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:58885201 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATAACTATGGAAAACAGAGTAGC[A/G]AACACAAAGAAACAGAAACTGTAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLTM PubMed Links

Gene Details

Gene
SLTM
Gene Name
SAFB like transcription modulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013843.2 Intron NP_001013865.1
NM_024755.3 Intron NP_079031.2
XM_006720686.3 Intron XP_006720749.3
XM_006720690.1 Intron XP_006720753.1
XM_011522022.1 Intron XP_011520324.1
XM_011522023.1 Intron XP_011520325.1
XM_011522024.1 Intron XP_011520326.1
XM_011522025.1 Intron XP_011520327.1
XM_011522026.2 Intron XP_011520328.2
XM_011522027.1 Intron XP_011520329.1
XM_011522028.1 Intron XP_011520330.1
XM_011522029.2 Intron XP_011520331.1
XM_011522030.2 Intron XP_011520332.1
XM_011522031.2 Intron XP_011520333.1
XM_017022576.1 Intron XP_016878065.1
XM_017022577.1 Intron XP_016878066.1
XM_017022578.1 Intron XP_016878067.1
XM_017022579.1 Intron XP_016878068.1
XM_017022580.1 Intron XP_016878069.1

View Full Product Details