Product Details
- SNP ID
-
rs7619492
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:111048981 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAACTTTTACTATTATAAGCAACAT[C/T]GCAATGAATATTTGTGTACATTATT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PVRL3-AS1
PubMed Links
Gene Details
- Gene
- PVRL3-AS1
- Gene Name
- PVRL3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details