Product Details

SNP ID
rs7161110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:57995539 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAAGTTAAATCAAATGTAAAAAA[C/T]GTGTGCCCTATAATAATAAATGACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C14orf37 PubMed Links

Gene Details

Gene
C14orf37
Gene Name
chromosome 14 open reading frame 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001872.3 11600 Intron NP_001001872.2
NM_001320173.1 11600 Intron NP_001307102.1
XM_011536471.2 11600 Intron XP_011534773.1
XM_017021029.1 11600 UTR 3 XP_016876518.1
XM_017021030.1 11600 UTR 3 XP_016876519.1

View Full Product Details