Product Details

SNP ID
rs4598831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:91785019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAGCAAAAAGTCTGACAAGTTT[C/T]GATTTTTGTAGCAATTGAATTTTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TC2N PubMed Links

Gene Details

Gene
TC2N
Gene Name
tandem C2 domains, nuclear
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128595.2 Intron NP_001122067.1
NM_001128596.2 Intron NP_001122068.1
NM_001289134.1 Intron NP_001276063.1
NM_152332.5 Intron NP_689545.1

View Full Product Details