Product Details

SNP ID
rs7808288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:882921 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGCAGCTGTCCCCCTGTCTGCCC[A/G]CACGGTCGTGGGTCTGGCTCTTCCT
Phenotype
MIM: 612056 MIM: 607723
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GET4 PubMed Links

Gene Details

Gene
GET4
Gene Name
golgi to ER traffic protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015949.2 Intron NP_057033.2
Gene
SUN1
Gene Name
Sad1 and UNC84 domain containing 1
There are no transcripts associated with this gene.

View Full Product Details