Product Details

SNP ID
rs6824123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:13300141 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTGGTTTCTTACTGACACAAGAG[C/G]CTCCCAGAAAGGGGCACTGAAATAT
Phenotype
MIM: 612994
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RAB28 PubMed Links

Gene Details

Gene
RAB28
Gene Name
RAB28, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017979.2 Intron NP_001017979.1
NM_001159601.1 Intron NP_001153073.1
NM_004249.3 Intron NP_004240.2
XM_005248218.4 Intron XP_005248275.1
XM_011513911.2 Intron XP_011512213.1
XM_011513912.2 Intron XP_011512214.1
XM_017008850.1 Intron XP_016864339.1

View Full Product Details