Product Details

SNP ID
rs6954451
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:30137479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCCTTTCTCTTGAGGCCACCCA[G/T]ATTTTAAATATTTTCCCATCTGCAT
Phenotype
MIM: 608639
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MTURN PubMed Links

Gene Details

Gene
MTURN
Gene Name
maturin, neural progenitor differentiation regulator homolog (Xenopus)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152793.2 Intron NP_690006.2
XM_005249652.3 Intron XP_005249709.1
Gene
PLEKHA8
Gene Name
pleckstrin homology domain containing A8
There are no transcripts associated with this gene.

View Full Product Details