Product Details

SNP ID
rs6545484
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:55283609 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGTTCTTATTGCCTCCCCACTCAC[C/G]TTCTTTGTGGTTACATATTTGTAGA
Phenotype
MIM: 609736
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCDC88A PubMed Links

Gene Details

Gene
CCDC88A
Gene Name
coiled-coil domain containing 88A
There are no transcripts associated with this gene.

Gene
PRORSD1P
Gene Name
prolyl-tRNA synthetase associated domain containing 1, pseudogene
There are no transcripts associated with this gene.

View Full Product Details