Product Details

SNP ID
rs7741300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32046212 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCAGAGACCGGGCCCAGGCGCTT[C/T]CCCCCAAGGAGCCCGTAGAGCAGAA
Phenotype
MIM: 613815 MIM: 600985
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP21A2 PubMed Links

Gene Details

Gene
CYP21A2
Gene Name
cytochrome P450 family 21 subfamily A member 2
There are no transcripts associated with this gene.

Gene
TNXB
Gene Name
tenascin XB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019105.6 10765 Silent Mutation GGA,GGG G3521G NP_061978.6
NM_032470.3 10765 Intron NP_115859.2

View Full Product Details