Product Details

SNP ID
rs6902650
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:41796342 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCTTTTGTTTTTAGGAAAGGAGG[A/G]AACTCCCAAACTCATTCAAAAGAAA
Phenotype
MIM: 611389 MIM: 616168
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PRICKLE4 PubMed Links

Gene Details

Gene
PRICKLE4
Gene Name
prickle planar cell polarity protein 4
There are no transcripts associated with this gene.

Gene
TOMM6
Gene Name
translocase of outer mitochondrial membrane 6
There are no transcripts associated with this gene.

Gene
USP49
Gene Name
ubiquitin specific peptidase 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286554.1 2360 UTR 3 NP_001273483.1
NM_018561.4 2360 Intron NP_061031.2

View Full Product Details