Product Details

SNP ID
rs6932918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:45901322 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTCAGGCTTATAAAGTGTCCT[A/C]CTGAGAAAGTTCCTCTTCCTCAATG
Phenotype
MIM: 607293
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CLIC5 PubMed Links

Gene Details

Gene
CLIC5
Gene Name
chloride intracellular channel 5
There are no transcripts associated with this gene.

View Full Product Details