Product Details

SNP ID
rs6536886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:165032103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATCAGGCCTGAGCCGCCGGTCCA[A/G]CTGCTCCAGGTAAGCTGGGAGGGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM60 PubMed Links

Gene Details

Gene
TRIM60
Gene Name
tripartite motif containing 60
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258025.1 151 UTR 5 NP_001244954.1
NM_152620.2 151 UTR 5 NP_689833.1
XM_011531683.2 151 UTR 5 XP_011529985.1

View Full Product Details