Product Details

SNP ID
rs7997441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:28306203 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCTTTGTGGAGAAATTGTCAAGT[A/G]AGAAAAGGGTGCCAGGATGAAATGG
Phenotype
MIM: 165070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLT1 PubMed Links

Gene Details

Gene
FLT1
Gene Name
fms related tyrosine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159920.1 Intron NP_001153392.1
NM_001160030.1 Intron NP_001153502.1
NM_001160031.1 Intron NP_001153503.1
NM_002019.4 Intron NP_002010.2
XM_011535014.1 Intron XP_011533316.1
XM_017020485.1 Intron XP_016875974.1

View Full Product Details