Product Details

SNP ID
rs7989622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:38966187 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCATGTGCCTTTTCCCACCCTCT[A/G]ATAATGGTGCTGGAAGGTGGAGAGT
Phenotype
MIM: 608327
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STOML3 PubMed Links

Gene Details

Gene
STOML3
Gene Name
stomatin like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144033.1 1736 UTR 3 NP_001137505.1
NM_145286.2 1736 UTR 3 NP_660329.1
XM_017020400.1 1736 UTR 3 XP_016875889.1

View Full Product Details