Product Details

SNP ID
rs7766270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:127279520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACCAAACTGTCTTGTTACCTTA[A/G]CTTTGTAGTAAGTTTTGACATTGGA
Phenotype
MIM: 612136 MIM: 612137
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ECHDC1 PubMed Links

Gene Details

Gene
ECHDC1
Gene Name
ethylmalonyl-CoA decarboxylase 1
There are no transcripts associated with this gene.

Gene
RNF146
Gene Name
ring finger protein 146
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242844.1 Intron NP_001229773.1
NM_001242845.1 Intron NP_001229774.1
NM_001242846.1 Intron NP_001229775.1
NM_001242847.1 Intron NP_001229776.1
NM_001242848.1 Intron NP_001229777.1
NM_001242849.1 Intron NP_001229778.1
NM_001242850.1 Intron NP_001229779.1
NM_001242851.1 Intron NP_001229780.1
NM_001242852.1 Intron NP_001229781.1
NM_030963.3 Intron NP_112225.2
XM_006715571.3 Intron XP_006715634.1
XM_011536161.2 Intron XP_011534463.1
XM_011536162.2 Intron XP_011534464.1
XM_011536163.2 Intron XP_011534465.1
XM_011536164.2 Intron XP_011534466.1
XM_017011336.1 Intron XP_016866825.1
XM_017011337.1 Intron XP_016866826.1
XM_017011338.1 Intron XP_016866827.1
XM_017011339.1 Intron XP_016866828.1
XM_017011340.1 Intron XP_016866829.1
XM_017011341.1 Intron XP_016866830.1
XM_017011342.1 Intron XP_016866831.1
XM_017011343.1 Intron XP_016866832.1

View Full Product Details