Product Details

SNP ID
rs4464357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:45852008 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCACAGGTACAAAGTTAAAGTTA[A/G]ACAGGAGGAGTAAATTCTGGTGTTC
Phenotype
MIM: 608502
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCBP3 PubMed Links

Gene Details

Gene
PCBP3
Gene Name
poly(rC) binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130141.1 Intron NP_001123613.1
NM_020528.2 Intron NP_065389.2

View Full Product Details