Product Details

SNP ID
rs6516724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:14278181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAATGGGAAATAGATTCCACCTCT[C/T]GATGGGAAAAGCTGCAAAGAATTGG
Phenotype
MIM: 608835
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ABCC13 PubMed Links

Gene Details

Gene
ABCC13
Gene Name
ATP binding cassette subfamily C member 13 (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details