Product Details
- SNP ID
-
rs7509384
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:21708481 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGACCCCTGGCCGTGTCTTCAGATA[C/G]AAATTGGGTGGTTGGCCACACGTGT
- Phenotype
-
MIM: 167411
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101929608
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116287563] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC101929608
- Gene Name
- uncharacterized LOC101929608
There are no transcripts associated with this gene.
- Gene
- PAX1
- Gene Name
- paired box 1
View Full Product Details