Product Details

SNP ID
rs7569854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:183122927 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAGGTATTTTGAAGATGTGATTA[C/T]TAACTTAATCACCTCTACTGAATAT
Phenotype
MIM: 608140
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUP35 PubMed Links

Gene Details

Gene
NUP35
Gene Name
nucleoporin 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287584.1 Intron NP_001274513.1
NM_001287585.1 Intron NP_001274514.1
NM_138285.4 Intron NP_612142.2
XM_006712254.2 Intron XP_006712317.1
XM_011510576.2 Intron XP_011508878.1
XM_011510577.2 Intron XP_011508879.1
XM_017003308.1 Intron XP_016858797.1

View Full Product Details