Product Details

SNP ID
rs7061117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:133557989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTTTTTTTTTCTCGATTGATTT[C/G]TCCCAAGTGGTTCTCCCTATGCCAT
Phenotype
MIM: 300037
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GPC3 PubMed Links

Gene Details

Gene
GPC3
Gene Name
glypican 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164617.1 Intron NP_001158089.1
NM_001164618.1 Intron NP_001158090.1
NM_001164619.1 Intron NP_001158091.1
NM_004484.3 Intron NP_004475.1
XM_017029413.1 Intron XP_016884902.1

View Full Product Details